• USA Home
  • 22667 - 3-Quinolinecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
22667 Sigma-Aldrich

3-Quinolinecarboxylic acid

purum, ≥98.0% (HPLC)

  • CAS Number 6480-68-8

  • Empirical Formula (Hill Notation) C10H7NO2

  • Molecular Weight 173.17

  •  Beilstein Registry Number 126542

  •  EC Number 229-337-3

  •  MDL number MFCD00006770

  •  PubChem Substance ID 329752333

Purchase

Properties

Related Categories Building Blocks, C8 to C10, Chemical Synthesis, Heterocyclic Building Blocks, Quinolines More...
InChI Key   DJXNJVFEFSWHLY-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   277-280 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

177148 3-Quinolinecarboxylic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?