• USA Home
  • 22687 - Quinoline N-oxide hydrate

EMAIL THIS PAGE TO A FRIEND
22687 Sigma-Aldrich

Quinoline N-oxide hydrate

purum, ≥97.0% (HPLC)

  • CAS Number 64201-64-5

  • Empirical Formula (Hill Notation) C9H7NO · xH2O

  • Molecular Weight 145.16 (anhydrous basis)

  •  Beilstein Registry Number 5989589

  •  MDL number MFCD00149472

  •  PubChem Substance ID 329752336

Purchase

Properties

InChI Key   CUSWDTBBCIXCRB-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (HPLC)
impurities   ~20% water
mp   ~50 °C
  52-55 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

122327 Quinoline N-oxide hydrate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?