• USA Home
  • 22718 - Chitin from shrimp shells

EMAIL THIS PAGE TO A FRIEND
22718 Sigma-Aldrich

Chitin from shrimp shells

Synonym: Poly-(1→4)-β-N-acetyl-D-glucosamine

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates A-C, Polysaccharide More...
InChI Key   OVRNDRQMDRJTHS-WTZNIHQSSA-N
suitability   suitable as substrate for chitinase test

Description

Other Notes

Review of properties and possible applications; In immobilization techniques. Fructose production by immobilized inulinase; In the bacterial production of N,N′-diacetylchitobiose; Preparation of chitin gels

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?