• USA Home
  • 228737 - Zinc nitrate hexahydrate

EMAIL THIS PAGE TO A FRIEND
228737 Sigma-Aldrich

Zinc nitrate hexahydrate

reagent grade, 98%

Purchase

Properties

Related Categories Essential Chemicals, Research Essentials, Salts, Zinc Salts
grade   reagent grade
assay   98%
solubility   water: soluble(lit.)

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

100, 500 g in poly bottle

7 kg in poly drum

Application

Used to synthesize a new microporous, organically templated zinc phosphate. Such compounds have potential applications as catalysts or in other applications involving microporosity.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1514 5.1 / PGII
WGK Germany 
3
RTECS 
ZH4775000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Selected Applications of Metal-Organic Frameworks in Sustainable Energy Technologies

Although the field of high surface area materials is considered very mature, the need for improved materials for alternative energy applications remains pressing. Currently, zeolites, metal oxides, a...
Stephen R. Caskey, Adam J. Matzger
Material Matters 2009, 4.4, 111.
Keywords: Adsorption, Alternative energy, Applications, Capabilities, Combustion, Diffraction, Environmental, Filtration, Infrared spectroscopy, Ligands, Metal organic frameworks, Purification, Scintillation, Separation, Solvents, Sonication, Spectra, Tools, Type, X-Ray diffraction

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?