• USA Home
  • 22990 - 2-Chloroacrylonitrile

EMAIL THIS PAGE TO A FRIEND
22990 Sigma-Aldrich

2-Chloroacrylonitrile

purum, ≥97.0% (GC)

  • CAS Number 920-37-6

  • Linear Formula CH2=CClCN

  • Molecular Weight 87.51

  •  Beilstein Registry Number 773744

  •  MDL number MFCD00001858

  •  PubChem Substance ID 329752361

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   OYUNTGBISCIYPW-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
impurities   ≤1% acrylonitrile
refractive index   n20/D 1.429(lit.)
  n20/D 1.431
bp   88-89 °C(lit.)
mp   −65 °C(lit.)
density   1.096 g/mL at 25 °C(lit.)

Description

Other Notes

A ketene equivalent. Review

General description

The attachment of electron to neutral clusters of 2-chloroacrylonitrile (CAN) was studied.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
RIDADR 
UN 3383 6.1(3) / PGI
WGK Germany 
3
Flash Point(F) 
42.8 °F
Flash Point(C) 
6 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C22369 2-Chloroacrylonitrile, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?