• USA Home
  • 230154 - Resorufin sodium salt

EMAIL THIS PAGE TO A FRIEND
230154 Sigma-Aldrich

Resorufin sodium salt

Dye content 75 %

Synonym: 7-Hydroxy-3H-phenoxazin-3-one sodium salt

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Q-R, Stains & Dyes, A to Z, Stains and Dyes More...
description   Redox indicator
InChI Key   TYXBZZBYMSVFPR-UHFFFAOYSA-M
composition   Dye content, 75%
mp   >300 °C(lit.)
absorption   λmax 573 nm

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

R3257 Resorufin sodium salt, redox indicator

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?