• USA Home
  • 23015 - Sodium 2-chloroethanesulfonate monohydrate

EMAIL THIS PAGE TO A FRIEND
23015 Sigma-Aldrich

Sodium 2-chloroethanesulfonate monohydrate

purum, ≥97.0% (AT)

Synonym: 2-Chloroethanesulfonic acid sodium salt monohydrate

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acid Salts, Sulfur Compounds More...
InChI Key   OSEJMOGPHHVJJG-UHFFFAOYSA-M
grade   purum
assay   ≥97.0% (AT)
mp   292 °C (dec.)(lit.)
solubility   H2O: soluble0.1 g/mL, clear

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?