Synonym: Epsom salts
CAS Number 10034-99-8
Linear Formula MgSO4 · 7H2O
Molecular Weight 246.47
EC Number 231-298-2
MDL number MFCD00149785
PubChem Substance ID 24853801
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | ACS Grade, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| vapor density | <0.01 (vs air) |
| vapor pressure | <0.1 mmHg ( 20 °C) |
| assay | ≥98% |
| 98.0-102.0% (ACS specification) | |
| impurities | ≤0.005% insolubles |
| pH | 5.0-8.2 (25 °C, 5%) |
1, 2.5 kg in poly bottle
5 kg in poly drum
12 kg in fiber drum
25, 500 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?