• USA Home
  • 23040 - 2-Chloroethyl isocyanate

EMAIL THIS PAGE TO A FRIEND
23040 Sigma-Aldrich

2-Chloroethyl isocyanate

technical, ≥95% (GC)

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanates, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   BCMYXYHEMGPZJN-UHFFFAOYSA-N
grade   technical
assay   ≥95% (GC)
impurities   1,2-dichloroethane
refractive index   n20/D 1.447(lit.)
  n20/D 1.447
bp   141-142 °C(lit.)
density   1.237 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 2478PSN1 6.1(3) / PGIII
WGK Germany 
3
RTECS 
NQ8450000
Flash Point(F) 
132.8 °F
Flash Point(C) 
56 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

538337 2-Chloroethyl isocyanate, low HCl, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?