• USA Home
  • 23050 - 2-Chloroethyl methyl sulfide

EMAIL THIS PAGE TO A FRIEND
23050 Sigma-Aldrich

2-Chloroethyl methyl sulfide

purum, ≥97.0% (GC)

Synonym: 2-(Methylthio)ethyl chloride, Hemisulfur mustard

  • CAS Number 542-81-4

  • Linear Formula ClCH2CH2SCH3

  • Molecular Weight 110.61

  •  Beilstein Registry Number 1731033

  •  EC Number 208-828-6

  •  MDL number MFCD00000978

  •  PubChem Substance ID 329752374

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfides/Disulfides, Sulfur Compounds More...
InChI Key   MYFKLQFBFSHBPA-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.496(lit.)
  n20/D 1.496
bp   55-56 °C/30 mmHg(lit.)
density   1.11 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Caution

may discolor to greenish-yellow on storage

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2929 3(6.1) / PGII
WGK Germany 
3
Flash Point(F) 
107.6 °F
Flash Point(C) 
42 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

242632 2-Chloroethyl methyl sulfide, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?