EMAIL THIS PAGE TO A FRIEND
23125 Sigma-Aldrich

Chlorambucil

purum, ≥98.0% (T)

Synonym: 4-(4-[Bis(2-chloroethyl)amino]phenyl)butyric acid, 4-[Bis(2-chloroethyl)amino]benzenebutyric acid

  • CAS Number 305-03-3

  • Empirical Formula (Hill Notation) C14H19Cl2NO2

  • Molecular Weight 304.21

  •  Beilstein Registry Number 999011

  •  EC Number 206-162-0

  •  MDL number MFCD00021783

  •  PubChem Substance ID 329752386

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Other Unnatural Amino Acids & Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   JCKYGMPEJWAADB-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   65-70 °C
storage temp.   2-8°C
Gene Information   human ... CYP2D6(1565)

Description

Other Notes

Alkylating antitumor agent, can be covalently coupled to monoclonal anticarcinoembryonic antigen

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
ES7525000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?