EMAIL THIS PAGE TO A FRIEND
23175 Sigma-Aldrich

Butyl chloroformate

purum, ≥97.0% (GC)

  • CAS Number 592-34-7

  • Linear Formula ClCOOCH2CH2CH2CH3

  • Molecular Weight 136.58

  •  Beilstein Registry Number 605635

  •  EC Number 209-750-5

  •  MDL number MFCD00000650

  •  PubChem Substance ID 329752389

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
vapor pressure   2.42 psi ( 20 °C)
InChI Key   NRDQFWXVTPZZAZ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.412(lit.)
  n20/D 1.413
bp   142 °C(lit.)
density   1.057 g/mL at 20 °C
  1.074 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2743 - class 6.1 - PG 1 - n-Butyl chloroformate
WGK Germany 
1
Flash Point(F) 
100.4 °F
Flash Point(C) 
38 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

184462 Butyl chloroformate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?