• USA Home
  • 23253 - Propargyl chloroformate

EMAIL THIS PAGE TO A FRIEND
23253 Sigma-Aldrich

Propargyl chloroformate

technical, ≥90% (GC)

  • CAS Number 35718-08-2

  • Linear Formula HC≡CCH2OCOCl

  • Molecular Weight 118.52

  •  Beilstein Registry Number 1745399

  •  MDL number MFCD00270127

  •  PubChem Substance ID 329752401

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
vapor pressure   0.65 psi ( 20 °C)
InChI Key   RAMTXCRMKBFPRG-UHFFFAOYSA-N
grade   technical
assay   ≥90% (GC)
refractive index   n20/D 1.435(lit.)
  n20/D 1.435
bp   118-122 °C(lit.)
density   1.215 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3390 6.1/PG 1
WGK Germany 
3
Flash Point(F) 
86 °F
Flash Point(C) 
30 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?