• USA Home
  • 23255 - 2-(1-Pyrenyl)ethyl chloroformate

EMAIL THIS PAGE TO A FRIEND
23255 Sigma-Aldrich

2-(1-Pyrenyl)ethyl chloroformate

for fluorescence, ≥97.0% (HPLC)

Purchase

Properties

Related Categories Biochemicals and Reagents, Derivatization agents, Fluorescent Probes, Labels, Particles and Stains, Functional Group Reactive Labels, Functional Group Reactive Labels for Chromatography,
grade   for fluorescence
InChI Key   VEQBZYDZNNCKFN-UHFFFAOYSA-N
assay   ≥97.0% (HPLC)
mp   68-72 °C
fluorescence   λex 326 nm; λem 376 nm in acetonitrile
  λex 326 nm; λem 396 nm in acetonitrile
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?