• USA Home
  • 23271 - Chloramine T polymer-bound

EMAIL THIS PAGE TO A FRIEND
23271 Sigma-Aldrich

Chloramine T polymer-bound

beads, extent of labeling: 0.20 mmol/g chloramine T loading

Synonym: Iodo-Beads®

Purchase

Properties

Related Categories Chemical Synthesis, Oxidizing Reagents, Solid Supported Synthesis, Specialty Synthesis, Supported Reagents More...
form   beads
extent of labeling   0.20 mmol/g chloramine T loading
diam.   ~3 mm
capacity   0.55 μmol/bead±0.05 μmol/bead oxidative capacity
storage temp.   2-8°C

Description

General description

The product consists of N-chloro-benzenesulfonamide immobilised on 2.8 mm-diameter non-porous polystyrene spheres.

Other Notes

Solid-state reagent for iodinating peptides and proteins. It consists of N-chloro-benzenesulfonamide immobilised on 2.8 mm-diameter non-porous polystyrene spheres

Packaging

Package of 50 beads

Legal Information

Iodo-Beads is a registered trademark of Pierce Biotechnology, Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?