EMAIL THIS PAGE TO A FRIEND
23350 Sigma-Aldrich

4-Chloroaniline

technical, ≥97.0% (GC)

  • CAS Number 106-47-8

  • Linear Formula ClC6H4NH2

  • Molecular Weight 127.57

  •  Beilstein Registry Number 471359

  •  EC Number 203-401-0

  •  MDL number MFCD00007835

Purchase

Properties

vapor density   4.4 (vs air)
vapor pressure   0.15 mmHg ( 25 °C)
InChI Key   QSNSCYSYFYORTR-UHFFFAOYSA-N
grade   technical
assay   ≥97.0% (GC)
form   crystalline
  crystals
  crystals or chunks
bp   232 °C(lit.)
mp   67-70 °C(lit.)
  67-70 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2018 6.1 / PGII
WGK Germany 
3
RTECS 
BX0700000
Flash Point(F) 
248 °F
Flash Point(C) 
120 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

C22415 4-Chloroaniline, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?