CAS Number 7784-13-6
Empirical Formula (Hill Notation) AlCl3 · 6H2O
Molecular Weight 241.43
EC Number 231-208-1
MDL number MFCD00149134
PubChem Substance ID 24854214
Popular Documents: Specification Sheet (PDF)
| Related Categories | Aluminum, Catalysis and Inorganic Chemistry, Chemical Synthesis, Lewis Acids, Synthetic Reagents More... |
| grade | ReagentPlus® |
| vapor pressure | 1 mmHg ( 100 °C) |
| assay | 99% |
100, 500 g in poly bottle
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?