Synonym: Alum, Potassium alum, Potassium aluminum sulfate dodecahydrate
CAS Number 7784-24-9
Linear Formula AlK(SO4)2 · 12H2O
Molecular Weight 474.39
EC Number 233-141-3
MDL number MFCD00149143
PubChem Substance ID 24854215
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Aluminum, Catalysis and Inorganic Chemistry, Cell Culture, Chemical Synthesis, |
| grade | ACS reagent |
| assay | ≥98% |
| 98.0-102.0% (ACS specification) | |
| impurities | ≤0.005% insolubles |
| mp | 92 °C(lit.) |
| density | 1.757 g/mL at 25 °C(lit.) |
| anion traces | chloride (Cl-): ≤5 ppm |
100, 500 g in poly bottle
2.5 kg in poly bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?