237132 Sigma-Aldrich

Calcium sulfate

−325 mesh, 99%

DOWNLOAD MSDS (PDF)

Synonym: Gypsum

  • CAS Number 7778-18-9

  • Linear Formula CaSO4

  • Molecular Weight 136.14

  •  EC Number 231-900-3

  •  MDL number MFCD00010912

  •  PubChem Substance ID 24854220

Properties

assay   99%
particle size   −325 mesh

Description

Packaging

100, 500 g in poly bottle

Price and Availability

Customers Also Viewed

Calcium sulfate

≥99.99% trace metals basis

Calcium sulfate dihydrate

ReagentPlus®, ≥99%

Calcium sulfate dihydrate

BioXtra, ≥98.0%

Calcium sulfate dihydrate

ACS reagent, 98%

Calcium sulfate hemihydrate

purum, ≥97.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?