CAS Number 7632-00-0
Linear Formula NaNO2
Molecular Weight 69.00
EC Number 231-555-9
MDL number MFCD00011118
PubChem Substance ID 24854227
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| assay | ≥97.0% |
| autoignition temp. | 914 °F |
| impurities | ≤0.01% insolubles |
| mp | 271 °C(lit.) |
| anion traces | chloride (Cl-): ≤0.005% |
| sulfate (SO42-): ≤0.01% |
100, 500 g in poly bottle
2.5 kg in poly bottle
5 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?