CAS Number 7722-64-7
Empirical Formula (Hill Notation) KMnO4
Molecular Weight 158.03
EC Number 231-760-3
MDL number MFCD00011364
PubChem Substance ID 24854310
Popular Documents: Specification Sheet (PDF)
| Related Categories | Chemical Synthesis, Essential Chemicals, Inorganic Salts, Solutions and Reagents, Others, Oxidizing Agents, |
| assay | 97% |
| form | powder |
| particle size | −325 mesh |
Layered MnO2 with very promising characteristics for battery applications is produced by thermal decomposition of KMnO4.1
Suitable for heterogeneous oxidations.
100, 500 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?