CAS Number 7757-82-6
Linear Formula Na2SO4
Molecular Weight 142.04
EC Number 231-820-9
MDL number MFCD00003504
PubChem Substance ID 24854335
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Adsorbents, Filter Aids and Drying Agents, Chemical Synthesis, Essential Chemicals, Inorganic Salts, |
| grade | ACS reagent |
| anhydrous | |
| assay | ≥99.0% |
| form | powder |
| impurities | ≤0.01% insolubles |
| ≤5 ppm N compounds | |
| loss | ≤0.5% loss on ignition |
12, 25 kg in fiber drum
25, 500, 36×500 g in poly bottle
2.5 kg in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?