EMAIL THIS PAGE TO A FRIEND
238929 Sigma-Aldrich

Drierite

without indicator, 6 mesh

Synonym: Calcium sulfate, Gypsum

Purchase

Properties

Related Categories Adsorbents, Filter Aids and Drying Agents, Essential Chemicals, Other Drying Agents, Research Essentials
InChI Key   OSGAYBCDTDRGGQ-UHFFFAOYSA-L
particle size   6 mesh

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Legal Information

Drierite is a trademark of W.A. Hammond Drierite Co., LTD

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
WS6920000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?