EMAIL THIS PAGE TO A FRIEND
239186 Sigma-Aldrich

Aluminum hydroxide

reagent grade

Purchase

Properties

Related Categories Aluminum Salts, Cell Culture, Essential Chemicals, Mineral salt adjuvants, Research Essentials,
grade   reagent grade
composition   Al2O3, 50-57%

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

may contain aluminum carbonate species

Packaging

2 kg in fiber drum

25, 500 g in poly bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
BD0940000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?