CAS Number 1305-62-0
Linear Formula Ca(OH)2
Molecular Weight 74.09
EC Number 215-137-3
MDL number MFCD00010901
PubChem Substance ID 24854362
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Chemical Synthesis, Essential Chemicals, Inorganic Bases, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| assay | ≥95.0% |
| impurities | ≤0.03% insol. in HCl |
| ≤0.1% S compounds | |
| ≤3.0% calcium carbonate | |
| density | 2.24 g/mL at 25 °C(lit.) |
| anion traces | chloride (Cl-): ≤0.03% |
100, 500 g in poly bottle
2.5 kg in poly drum
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?