CAS Number 7789-02-8
Linear Formula Cr(NO3)3 · 9H2O
Molecular Weight 400.15
EC Number 236-921-1
MDL number MFCD00149659
PubChem Substance ID 24854364
Popular Documents: Specification Sheet (PDF)
| Related Categories | Catalysis and Inorganic Chemistry, Chemical Synthesis, Chromium More... |
| assay | 99% |
| mp | 60 °C(lit.) |
100, 500 g in poly bottle
2 kg in poly bottle
5 g in glass bottle
Customers Also Viewed
analytical standard
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?