-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym: Acetic acid sodium salt
CAS Number 127-09-3
Linear Formula CH3COONa
Molecular Weight 82.03
Beilstein Registry Number 3595639
EC Number 204-823-8
MDL number MFCD00012459
PubChem Substance ID 24854471
Popular Documents: Product Information Sheet (PDF)
| Related Categories | ACS Grade, ACS Grade Buffers, Analytical/Chromatography, Biological Buffers, Buffers A to Z, |
| grade | ACS reagent |
| assay | ≥99.0% |
| form | solid |
| autoignition temp. | 1112 °F |
| total impurities | ≤0.01% insolubles |
| loss | ≤1.0% loss on drying |
| pH | 7.0-9.2 (25 °C, 5%) |
1 kg in poly btl
5 g in glass btl
100, 500 g in poly btl
2.5 kg in poly drum
Whether you're looking for a specific brand, quality, or price, Sigma-Aldrich has the silica products to meet your unique purification or sample preparation requirements. Our new Silica Brochure makes it easy to find just the silica gel product you need.Customers Also Purchased
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?