24201 Sigma-Aldrich

Acetone

puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)

DOWNLOAD MSDS (PDF)
  • CAS Number 67-64-1

  • Linear Formula CH3COCH3

  • Molecular Weight 58.08

  •  Beilstein Registry Number 635680

  •  EC Number 200-662-2

  •  MDL number MFCD00008765

  •  PubChem Substance ID 24854528

Properties

Related Categories Acetone, Amber Glass Bottles, Multi-Compendial, Plastic Bottles, Solvent by Type,
vapor density   2 (vs air)
vapor pressure   184 mmHg ( 20 °C)
grade   puriss.
assay   ≥99% (GC)
quality   meets analytical specification of Ph. Eur., BP, NF
expl. lim.   13.2 %
impurities   acidity or alkalinity, complies
  reducing matter, complies
  related subst., complies (GC)
  residual solvents, complies
  water-insoluble matter, complies
  ≤0.0001% heavy metals (as Pb)
  ≤0.0002% benzene (GC)
  ≤0.002% non-volatile matter
  ≤0.05% 2-propanol (GC)
  ≤0.05% methanol (GC)
  ≤0.3% water (Karl Fischer)
refractive index   n20/D 1.359(lit.)
bp   56 °C/760 mmHg(lit.)
mp   −94 °C(lit.)
density   0.790-0.792 g/mL at 20 °C
  0.791 g/mL at 25 °C(lit.)
suitability   complies for appearance of solution
  complies for identity (IR)

Description

Packaging

2.5, 4×2.5 L-GL in glass bottle

Application

Acetone′s luminesence intensity is dependent upon the solution components 1. The absorption of UV light by acetone, results in its photolysis and the production of radials 1.

Price and Availability

Customers Also Viewed

2-Propanol

puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)

Acetone
34850Acetone

CHROMASOLV®, for HPLC, ≥99.8%

Acetone
179124Acetone

ACS reagent, ≥99.5%

Acetone
32201Acetone

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)

Acetone
00585Acetone

purum, ≥99.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?