Synonym: Amidosulfonic acid
CAS Number 5329-14-6
Linear Formula NH2SO3H
Molecular Weight 97.09
EC Number 226-218-8
MDL number MFCD00011603
PubChem Substance ID 24854581
Popular Documents: Specification Sheet (PDF)
| Related Categories | Bioactive Small Molecules, Cell Biology, Essential Chemicals, Inorganic Salts, Solutions and Reagents, Reagent Plus, |
| grade | ReagentPlus® |
| assay | ≥99% |
| mp | 215-225 °C (dec.)(lit.) |
| Gene Information | human ... CA1(759), CA2(760) |
100, 500 g in poly bottle
5 g in glass bottle
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?