EMAIL THIS PAGE TO A FRIEND
243477 Sigma-Aldrich

Zinc

ACS reagent, −30-+100 mesh, ≥99.8%, granular

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Metal and Ceramic Science, Metals,
grade   ACS reagent
vapor pressure   1 mmHg ( 487 °C)
InChI Key   HCHKCACWOHOZIP-UHFFFAOYSA-N
assay   ≥99.8%
form   granular
resistivity   5.8 μΩ-cm, 20°C
particle size   −30-+100 mesh
bp   907 °C(lit.)
mp   420 °C(lit.)
density   7.133 g/mL at 25 °C(lit.)
cation traces   Fe: ≤0.01%
  Pb: ≤0.01%
suitability   passes test for determination of arsenic (As)

Price and Availability


Laboratory Gloves

Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?