EMAIL THIS PAGE TO A FRIEND
243604 Sigma-Aldrich

Talc

powder, 10 μm

Synonym: Hydrous magnesium silicate

Purchase

Properties

InChI Key   FPAFDBFIGPHWGO-UHFFFAOYSA-N
form   powder
particle size   10 μm

Description

Packaging

2.5 kg in poly drum

500 g in poly bottle

Application

Talc (Hydrous magnesium silicate) may be used to compose the leached particle medium for the production of 2-phenylethanol by using Aspergillus niger DSM 821 by the microparticle-enhanced cultivation (MPEC).

General description

Microscopic analysis of talc suggests that it has a heterogeneously splintered structure.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
WW2710000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?