CAS Number 10035-10-6
Empirical Formula (Hill Notation) HBr
Molecular Weight 80.91
Beilstein Registry Number 3587158
MDL number MFCD00011323
PubChem Substance ID 24854661
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, ACS Grade Acids & Bases, Acids & Bases, Analytical Reagents, Analytical Reagents for General Use, |
| grade | ACS reagent |
| vapor density | 2.8 (vs air) |
| vapor pressure | 320 psi ( 21.1 °C) |
| 8 mmHg ( 25 °C) | |
| description | 48% aqueous HBr |
| assay | 47.0-49.0% (ACS specification) |
| 48% |
ACS specifications same as product 438065
1 L in glass bottle
100, 500 mL in glass bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?