EMAIL THIS PAGE TO A FRIEND
24520 Sigma-Aldrich

Chloroacetic acid

purum, ≥97.0% (T)

Synonym: Monochloroacetic acid

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Organic Acids, Synthetic Reagents More...
vapor density   3.26 (vs air)
vapor pressure   0.75 mmHg ( 20 °C)
InChI Key   FOCAUTSVDIKZOP-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (T)
ign. residue   ≤0.1%
bp   189 °C(lit.)
mp   60-63 °C(lit.)
  61-64 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1751 8(6.1) / PGII
WGK Germany 
3
RTECS 
AF8575000
Flash Point(F) 
258.8 °F
Flash Point(C) 
126 °C
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?