EMAIL THIS PAGE TO A FRIEND
24529 Sigma-Aldrich

Toluene

puriss., ≥99.5% (GC)

  • CAS Number 108-88-3

  • Linear Formula C6H5CH3

  • Molecular Weight 92.14

  •  Beilstein Registry Number 635760

  •  EC Number 203-625-9

  •  MDL number MFCD00008512

  •  eCl@ss 39011102

Purchase

Properties

Related Categories Aluminum Bottles, Amber Glass Bottles, Reagent, Solvent Bottles, Solvent Packaging Options,
vapor density   3.2 (vs air)
vapor pressure   22 mmHg ( 20 °C)
  26 mmHg ( 25 °C)
InChI Key   YXFVVABEGXRONW-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.5% (GC)
autoignition temp.   997 °F
expl. lim.   7 %
impurities   ≤0.0005% thiophene
  ≤0.002% non-volatile matter
  ≤0.05% water (Karl Fischer)
refractive index   n/D 1.496(lit.)
bp   110-111 °C(lit.)
mp   -93 °C(lit.)
density   0.865 g/mL at 25 °C(lit.)

Description

Other Notes

Pertains only in Germany: unversteuert

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Application

Toluene may be used for the sterilization of microbial cultures and lyse bacterial cells in the assay of bacterial enzymes. It may be used as solvent for the fabrication of high-resolution 3D structures of polylactide-based materials, via multi-photon polymerization. It was used for the activation of glass slides, for the in vitro test system for allergy diagnosis.

General description

Toluene, a benzene analog, is widely employed as an organic solvent. It has been reported to be a biotoxic solvent that kills most microorganisms at low concentrations (0.1%v/v). Its anaerobic degradation to CO2, by the denitrifying bacterium Thauera arornatica has been reported. It forms syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1294 - class 3 - PG 2 - Toluene
WGK Germany 
2
RTECS 
XS5250000
Flash Point(F) 
39.2 °F
Flash Point(C) 
4 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?