• USA Home
  • 24594 - Hamilton® syringe, 7000 series, knurled hub with chaney adapter
EMAIL THIS PAGE TO A FRIEND
24594 Sigma-Aldrich
Hamilton® syringe, 7000 series, knurled hub with chaney adapter
7000.5, volume 0.5 μL, needle size 25 ga (blunt tip), needle L 70 mm (2.75 in.)
Purchase
Properties
Related Categories Analytical/Chromatography, GC Manual Syringes, Hamilton 7000 Series, Knurled Hub with Chaney Adapter, Liquid sampling, Syringe with sample in the needle, 0.5 - 5 µL,
material   stainless steel needle blunt tip (point style 3)
needle size   25 ga (blunt tip)
feature   KH (Knurled Hub)
packaging   pkg of 1 ea
mfr. no.   Hamilton, 86252
needle L   70 mm (2.75 in.)
volume   0.5 μL
Description
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?