• USA Home
  • 24625 - 2-Chloro-6-fluorobenzonitrile

EMAIL THIS PAGE TO A FRIEND
24625 Sigma-Aldrich

2-Chloro-6-fluorobenzonitrile

purum, ≥98.0% (GC)

  • CAS Number 668-45-1

  • Linear Formula ClC6H3(F)CN

  • Molecular Weight 155.56

  •  Beilstein Registry Number 1940332

  •  EC Number 211-571-2

  •  MDL number MFCD00001780

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C7, Chemical Synthesis, Fluorinated Building Blocks,
InChI Key   XPTAYRHLHAFUOS-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   104 °C/11 mmHg(lit.)
mp   55-59 °C(lit.)
  55-59 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3439 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

188182 2-Chloro-6-fluorobenzonitrile, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?