246980 Sigma-Aldrich

Sodium sulfate decahydrate

reagent grade, 97%

DOWNLOAD MSDS (PDF)

Synonym: Glauber’s salt

  • CAS Number 7727-73-3

  • Linear Formula Na2SO4 · 10H2O

  • Molecular Weight 322.20

  •  EC Number 231-820-9

  •  MDL number MFCD00149189

  •  PubChem Substance ID 24854839

Properties

Related Categories Essential Chemicals, Inorganic Salts, Solutions and Reagents, Reagent Grade, Research Essentials
grade   reagent grade
assay   97%
density   1.46 g/mL at 25 °C(lit.)

Description

Packaging

2.5 kg in poly bottle

500 g in poly bottle

Price and Availability

Customers Also Viewed

Sodium sulfate

ACS reagent, ≥99.0%, anhydrous, granular

Sodium sulfate

ACS reagent, ≥99.0%, anhydrous, powder

Sodium sulfate decahydrate

ACS reagent, ≥99.0%

Sodium sulfate decahydrate

BioUltra, ≥99.0% (calc. on dry substance, T)

Sodium sulfate decahydrate

puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?