EMAIL THIS PAGE TO A FRIEND
24798 Sigma-Aldrich

Chlorhexidine

purum, ≥99.0% (HPLC)

Synonym: 1,1′-Hexamethylenebis[5-(4-chlorophenyl)biguanide]

  • CAS Number 55-56-1

  • Linear Formula [-(CH2)3NHC(=NH)NHC(=NH)NHC6H4Cl]2

  • Molecular Weight 505.45

  •  Beilstein Registry Number 2826432

  •  EC Number 200-238-7

  •  MDL number MFCD00009673

  •  PubChem Substance ID 24854905

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Guanidines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   GHXZTYHSJHQHIJ-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (HPLC)
mp   134-136 °C(lit.)
  134-136 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
DU1925000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

282227 Chlorhexidine, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?