• USA Home
  • 24840 - (R)-(+)-4-Chloro-3-hydroxybutyronitrile

EMAIL THIS PAGE TO A FRIEND
24840 Sigma-Aldrich

(R)-(+)-4-Chloro-3-hydroxybutyronitrile

purum, ≥97.0% (GC)

  • CAS Number 84367-31-7

  • Linear Formula ClCH2CH(OH)CH2CN

  • Molecular Weight 119.55

  •  Beilstein Registry Number 1700586

  •  MDL number MFCD00273362

Purchase

Properties

InChI Key   LHBPNZDUNCZWFL-SCSAIBSYSA-N
grade   purum
assay   ≥97.0% (GC)
optical activity   [α]20/D +19±2°, c = 2% in ethanol
refractive index   n20/D 1.474(lit.)
  n20/D 1.474
bp   110 °C/1 mmHg(lit.)
density   1.250 g/mL at 20 °C(lit.)

Description

Other Notes

Chiral building block; synthesis of R(-)-carnitine

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
235 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

456152 (R)-(+)-4-Chloro-3-hydroxybutyronitrile, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?