Synonym: Lime, Quicklime
CAS Number 1305-78-8
Linear Formula CaO
Molecular Weight 56.08
EC Number 215-138-9
MDL number MFCD00010911
PubChem Substance ID 24854936
Popular Documents: Specification Sheet (PDF)
| Related Categories | Calcium, Essential Chemicals, Inorganic Salts, Solutions and Reagents, Materials Science, Metal and Ceramic Science, |
| grade | reagent grade |
| form | powder |
| loss | 10% loss on ignition, 1000°C (absorbed H2O and CO2) |
| bp | 2850 °C(lit.) |
| density | 3.3 g/mL at 25 °C(lit.) |
Units are 10% in excess of stated weight to compensate for the loss of H2O and CO2
2.5 kg in poly drum
500 g in poly bottle
Customers Also Viewed

puriss., meets analytical specification of FCC, 96-100.5% (ex ignited substance), powder (fine)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?