• USA Home
  • 24888 - (3-Chloro-2-hydroxypropyl)trimethylammonium chloride solution

EMAIL THIS PAGE TO A FRIEND
24888 Sigma-Aldrich

(3-Chloro-2-hydroxypropyl)trimethylammonium chloride solution

purum, ~65% in H2O (T)

  • CAS Number 3327-22-8

  • Linear Formula ClCH2CH(OH)CH2N(Cl)(CH3)3

  • Molecular Weight 188.10

  •  Beilstein Registry Number 6576172

  •  EC Number 222-048-3

  •  MDL number MFCD00055655

  •  PubChem Substance ID 24854957

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   CSPHGSFZFWKVDL-UHFFFAOYSA-M
grade   purum
concentration   ~65% in H2O (T)
refractive index   n20/D 1.453

Description

Other Notes

Reagent for the cationization of cellulose, starch and related compounds

This product has been replaced by 348287-ALDRICH | (3-Chloro-2-hydroxypropyl)trimethylammonium chloride solution 60 wt. % in H2O

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
2
RTECS 
BP5275400
Flash Point(F) 
235 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?