• USA Home
  • 24899 - Chloride-selective liquid ion-exchanger microelectrode cocktail A

EMAIL THIS PAGE TO A FRIEND
24899 Sigma-Aldrich

Chloride-selective liquid ion-exchanger microelectrode cocktail A

Selectophore

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophore Cocktails More...
grade   Selectophore

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Cocktail used for the intracellular determination of chloride with a microelectrode; alkali metals as well as pH changes (4-9) do not interfere

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

98183 Chloride-selective liquid ion-exchanger microelectrode cocktail A, Selectophore

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?