• USA Home
  • 25060 - 3-Chloro-2-methylaniline

EMAIL THIS PAGE TO A FRIEND
25060 Sigma-Aldrich

3-Chloro-2-methylaniline

technical, ≥98.0% (GC)

Synonym: 2-Amino-6-chlorotoluene, 3-Chloro-o-toluidine

  • CAS Number 87-60-5

  • Linear Formula ClC6H3(CH3)NH2

  • Molecular Weight 141.60

  •  Beilstein Registry Number 471697

  •  EC Number 201-756-6

  •  MDL number MFCD00007766

  •  PubChem Substance ID 329752772

Purchase

Properties

InChI Key   ZUVPLKVDZNDZCM-UHFFFAOYSA-N
grade   technical
assay   ≥98.0% (GC)
refractive index   n20/D 1.588(lit.)
  n20/D 1.588
bp   115-117 °C/10 mmHg(lit.)
mp   2 °C(lit.)
density   1.185 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3429 6.1 / PGIII
WGK Germany 
2
RTECS 
XU4760000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

101621 3-Chloro-2-methylaniline, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?