• USA Home
  • 25080 - 4-Chloro-2-methylaniline

EMAIL THIS PAGE TO A FRIEND
25080 Sigma-Aldrich

4-Chloro-2-methylaniline

purum, ≥98.0% (GC)

Synonym: 2-Amino-5-chlorotoluene, 4-Chloro-o-toluidine

  • CAS Number 95-69-2

  • Linear Formula ClC6H3(CH3)NH2

  • Molecular Weight 141.60

  •  Beilstein Registry Number 878505

  •  EC Number 202-441-6

  •  MDL number MFCD00007842

  •  PubChem Substance ID 329752773

Purchase

Properties

InChI Key   CXNVOWPRHWWCQR-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.583(lit.)
bp   241 °C(lit.)
mp   24-27 °C(lit.)
  24-27 °C
density   1.19 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2239 6.1/PG 3
WGK Germany 
3
RTECS 
XU5000000
Flash Point(F) 
255 °F
Flash Point(C) 
124 °C
Protocols & Articles

Articles

GC Analysis of a 32-Component Anilines Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 32-Component Anilines Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography, Mass selective detector

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?