• USA Home
  • 25118 - (Chloromethyl)-isopropoxy-dimethylsilane

EMAIL THIS PAGE TO A FRIEND
25118 Sigma-Aldrich

(Chloromethyl)-isopropoxy-dimethylsilane

purum, ≥95.0% (GC)

Synonym: (Chloromethyl)dimethylisopropoxysilane

  • CAS Number 18171-11-4

  • Linear Formula ClCH2Si(CH3)2OCH(CH3)2

  • Molecular Weight 166.72

  •  Beilstein Registry Number 1736255

  •  MDL number MFCD00010605

Purchase

Properties

InChI Key   KOTFCAKXPLOYLN-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.415(lit.)
  n20/D 1.415
bp   64-66 °C/54 mmHg(lit.)
density   0.926 g/mL at 25 °C(lit.)

Description

Other Notes

Reagent for hydroxymethylations of carbonyl compounds via Grignard reaction and oxidative desilylation

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1993 3/PG 3
WGK Germany 
3
Flash Point(F) 
82 °F
Flash Point(C) 
28 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

292036 (Chloromethyl)-isopropoxy-dimethylsilane, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?