251402 Sigma-Aldrich

Trichloroethylene

ACS reagent, ≥99.5%

DOWNLOAD MSDS (PDF)

Synonym: TCE, Trichloroethene

Properties

Related Categories ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Cans with NPT Threads,
grade   ACS reagent
vapor density   4.5 (vs air)
vapor pressure   61 mmHg ( 20 °C)
assay   ≥99.5%
autoignition temp.   770 °F
impurities   Free halogens, passes test
  ≤0.0001 meq/g Titr. acid
  ≤0.0003 meq/g Titr. base
  ≤0.02% water
evapn. residue   ≤0.001%
color   APHA: ≤10
refractive index   n20/D 1.476(lit.)
bp   86.7 °C(lit.)
mp   −84.8 °C(lit.)
density   1.463 g/mL at 25 °C(lit.)
cation traces   heavy metals (as Pb): ≤1 ppm

Description

Packaging

1, 2, 2.5, 4×4 L in glass bottle

500 mL in glass bottle

Price and Availability

Customers Also Viewed

Tetrachloroethylene

anhydrous, ≥99%

Trichloroethylene

anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%

Trichloroethylene

spectrophotometric grade, ≥99.5%

Trichloroethylene

analytical standard, for environmental analysis

Trichloroethylene

puriss. p.a., ≥99.5% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?