• USA Home
  • 25260 - 2-(Chloromethyl)pyridine hydrochloride

EMAIL THIS PAGE TO A FRIEND
25260 Sigma-Aldrich

2-(Chloromethyl)pyridine hydrochloride

purum, ≥98.0% (AT)

Synonym: 2-Picolyl chloride hydrochloride

  • CAS Number 6959-47-3

  • Empirical Formula (Hill Notation) C6H6ClN · HCl

  • Molecular Weight 164.03

  •  Beilstein Registry Number 3689155

  •  EC Number 230-149-9

  •  MDL number MFCD00012811

  •  PubChem Substance ID 329752786

Purchase

Properties

Related Categories Building Blocks, C6, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
InChI Key   JPMRGPPMXHGKRO-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
mp   120-124 °C(lit.)
  120-124 °C

Description

Other Notes

This product has been replaced by 162701-ALDRICH | 2-(Chloromethyl)pyridine hydrochloride 98%

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGIII
WGK Germany 
3
RTECS 
US6825000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?