EMAIL THIS PAGE TO A FRIEND
25329 Sigma-Aldrich

4-Chloro-1-naphthol

≥95.0% (HPLC), tablet (~32 mg each)

  • CAS Number 604-44-4

  • Linear Formula ClC10H6OH

  • Molecular Weight 178.61

  •  Beilstein Registry Number 509750

  •  EC Number 210-068-5

  •  MDL number MFCD00003974

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Peroxidase Substrates, Substrates by Enzyme More...
InChI Key   LVSPDZAGCBEQAV-UHFFFAOYSA-N
assay   ≥95.0% (HPLC)
form   tablet (~32 mg each)
diam.   ~4 mm
mp   118-121 °C(lit.)
solubility   methanol: soluble1 tablet/10 mL, clear (≤2 min.)
suitability   in accordance for electrophoresis test
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?