CAS Number 10025-70-4
Linear Formula SrCl2 · 6H2O
Molecular Weight 266.62
EC Number 233-971-6
MDL number MFCD00149865
PubChem Substance ID 24855461
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| assay | 99% |
| 99.0-103.0% (ACS specification) | |
| impurities | ≤0.005% insolubles |
| pH | 5.0-7.0 (25 °C, 5%) |
| mp | 115 °C(lit.) |
| anion traces | sulfate (SO42-): ≤0.001% |
Used in the synthesis of cadmium sulfide core photocatalytic nanoparticles.1
100, 500 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?