CAS Number 10101-41-4
Linear Formula CaSO4 · 2H2O
Molecular Weight 172.17
EC Number 231-900-3
MDL number MFCD00149625
PubChem Substance ID 24855462
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Calcium, Chemical Synthesis, Essential Chemicals, Inorganic Salts, |
| grade | ACS reagent |
| assay | 98% |
| 98.0-102.0% (ACS specification) | |
| impurities | ≤0.02% insol. dil. HCl |
| anion traces | carbonate (CO32-): passes test |
| chloride (Cl-): ≤0.005% | |
| nitrate (NO3-): ≤0.005% |
1 kg in poly bottle
100, 500 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?